missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12444-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the complex I 75 kDa subunit family. Mammalian complex I is composed of 45 different subunits. It locates at the mitochondrial inner membrane. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone. This protein is the largest subunit of complex I and it is a component of the iron-sulfur fragment of the enzyme. It may form part of the active site crevice where NADH is oxidized. Mutations in this gene are associated with complex I deficiency. Several transcript variants encoding different isoforms have been found for this gene.Specifications
NDUFS1 | |
Polyclonal | |
Unconjugated | |
NDUFS1 | |
CI 75Kd, Complex I 75kD, NDUFS1, PRO1304 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
227197, 301458, 4719 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P28331, Q66HF1, Q91VD9 | |
NDUFS1 | |
NDUFS1 Fusion Protein Ag3135 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |