Learn More
NBAS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557831
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: AKGDLKFPLK IFQHSKPDLQ QKIIPDQDQL MAIALECIYT CERNDQLCLC YDLLECLPER GYGDKTEATT KLHDMVDQLE QILSVS Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.
NBAS Polyclonal antibody specifically detects NBAS in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
NBAS | |
Polyclonal | |
Unconjugated | |
NBAS | |
4933425L03Rik; ILFS2; NAG; NAG/BC035112 fusion; NAG/FAM49A fusion; NBAS; neuroblastoma amplified sequence; neuroblastoma-amplified gene protein; neuroblastoma-amplified protein; neuroblastoma-amplified protein-like; neuroblastoma-amplified sequence; similar to neuroblastoma-amplified protein; SOPH | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51594 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A2RRP1 | |
NBAS | |
Recombinant protein corresponding to Human NBAS | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only