Learn More
NAPE PLD Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555055
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: ELPVLKPYFI TNPEEAGVRE AGLRVTWLGH ATVMVEMDEL IFLTDPIFSS RASPSQYMGP KRFRRSPCTI SELPPIDAVL ISHNHYDHLD YNSV Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 93%.
NAPE PLD Polyclonal antibody specifically detects NAPE PLD in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
NAPE PLD | |
Polyclonal | |
Unconjugated | |
NAPEPLD | |
A530089G06; C7orf18; DKFZp781D1098; FMP30; Mbldc1; Metallo-beta-lactamase domain containing 1; N-acyl phosphatidylethanolamine phospholipase D; N-acyl-phosphatidylethanolamine-hydrolyzing phospholipase D; NAPE-hydrolyzing phospholipase D; NAPEPLD; NAPE-PLD | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
222236 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6IQ20 | |
NAPEPLD | |
Recombinant protein corresponding to Human NAPEPLD | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only