Learn More
n-Propyltrimethoxysilane, 98+%, Thermo Scientific Chemicals
£18.39 - £136.68
Chemical Identifiers
CAS | 1067-25-0 |
---|---|
Molecular Formula | C6H16O3Si |
Molecular Weight (g/mol) | 164.28 |
MDL Number | MFCD00043026 |
InChI Key | HQYALQRYBUJWDH-UHFFFAOYSA-N |
Synonym | trimethoxy propyl silane, n-propyltrimethoxysilane, propyltrimethoxysilane, silane, trimethoxypropyl, unii-t72c7n9xo2, dynasylan ptmo, trimethoxysilylpropane, propyl-trimethoxysilane, silane,trimethoxypropyl, trimethoxy-n-propylsilane |
PubChem CID | 61254 |
IUPAC Name | trimethoxy(propyl)silane |
SMILES | CCC[Si](OC)(OC)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11400127
|
Thermo Scientific Alfa Aesar
B21033.14 |
25 g |
£18.39
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11410127
|
Thermo Scientific Alfa Aesar
B21033.22 |
100 g |
£44.12
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11420127
|
Thermo Scientific Alfa Aesar
B21033.36 |
500 g |
£136.68
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It finds its application as an important material in sol-gel production, provides a hydrophobic surface treatment for inorganic powders or filter materials and it is one of the catalyst component for polyolefin production with Ziegler-Natta catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt finds its application as an important material in sol-gel production, provides a hydrophobic surface treatment for inorganic powders or filter materials and it is one of the catalyst component for polyolefin production with Ziegler-Natta catalyst.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
1067-25-0 | |
164.28 | |
HQYALQRYBUJWDH-UHFFFAOYSA-N | |
61254 | |
CCC[Si](OC)(OC)OC |
C6H16O3Si | |
MFCD00043026 | |
trimethoxy propyl silane, n-propyltrimethoxysilane, propyltrimethoxysilane, silane, trimethoxypropyl, unii-t72c7n9xo2, dynasylan ptmo, trimethoxysilylpropane, propyl-trimethoxysilane, silane,trimethoxypropyl, trimethoxy-n-propylsilane | |
trimethoxy(propyl)silane |
Specifications
142°C to 143°C | |
C6H16O3Si | |
UN1993 | |
34°C (93°F) | |
HQYALQRYBUJWDH-UHFFFAOYSA-N | |
trimethoxy(propyl)silane | |
61254 | |
≥98% |
1067-25-0 | |
MFCD00043026 | |
2070458 | |
trimethoxy propyl silane, n-propyltrimethoxysilane, propyltrimethoxysilane, silane, trimethoxypropyl, unii-t72c7n9xo2, dynasylan ptmo, trimethoxysilylpropane, propyl-trimethoxysilane, silane,trimethoxypropyl, trimethoxy-n-propylsilane | |
CCC[Si](OC)(OC)OC | |
164.28 | |
164.28 | |
n-Propyltrimethoxysilane |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'tsca' : TSCA
RUO – Research Use Only