missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylurethane, 98%, Thermo Scientific Chemicals

£36.60 - £104.00

Chemical Identifiers

CAS 101-99-5
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00026806
InChI Key LBKPGNUOUPTQKA-UHFFFAOYSA-N
Synonym n-phenylurethane, phenylurethane, ethyl phenylcarbamate, phenylurethan, ethyl carbanilate, euphorin, keimstop, urethane, phenyl, carbanilic acid, ethyl ester, phenylethyl carbamate
PubChem CID 7591
IUPAC Name ethyl N-phenylcarbamate
SMILES CCOC(=O)NC1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11341797
View Documents
Thermo Scientific Alfa Aesar
L05027.14
25 g
£36.60
25g
Please to purchase this item. Need a web account? Register with us today!
11351797
View Documents
Thermo Scientific Alfa Aesar
L05027.22
100 g
£104.00
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

101-99-5
165.192
LBKPGNUOUPTQKA-UHFFFAOYSA-N
7591
CCOC(=O)NC1=CC=CC=C1
C9H11NO2
MFCD00026806
n-phenylurethane, phenylurethane, ethyl phenylcarbamate, phenylurethan, ethyl carbanilate, euphorin, keimstop, urethane, phenyl, carbanilic acid, ethyl ester, phenylethyl carbamate
ethyl N-phenylcarbamate

Specifications

101-99-5
1.106
C9H11NO2
MFCD00026806
14,7320
LBKPGNUOUPTQKA-UHFFFAOYSA-N
ethyl N-phenylcarbamate
7591
98%
49°C to 51°C
237°C
1.5375
1942785
n-phenylurethane, phenylurethane, ethyl phenylcarbamate, phenylurethan, ethyl carbanilate, euphorin, keimstop, urethane, phenyl, carbanilic acid, ethyl ester, phenylethyl carbamate
CCOC(=O)NC1=CC=CC=C1
165.192
165.19
N-Phenylurethane
Videos
Safety and Handling

Safety and Handling

P201-P202-P281-P308+P313-P501c

H351

missing translation for 'einecsNumber' : 202-995-9

missing translation for 'rtecsNumber' : FD8925000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only