missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylformamide CHROMASOLV™ Plus, for HPLC, ≥99.9%, Honeywell Riedel-de Haën™
Solvent for many hydrophobic organic compounds.
£63.13 - £442.66
Chemical Identifiers
CAS | 68-12-2 |
---|---|
Molecular Formula | C3H7NO |
Molecular Weight (g/mol) | 73.10 |
MDL Number | MFCD00003284 |
InChI Key | ZMXDDKWLCZADIW-UHFFFAOYSA-N |
Synonym | dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa |
PubChem CID | 6228 |
ChEBI | CHEBI:17741 |
IUPAC Name | N,N-dimethylformamide |
SMILES | CN(C)C=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15652850
|
Honeywell-Chromasolv
270547-1L |
1 L |
£63.13
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15662850
|
Honeywell-Chromasolv
270547-2.5L |
2.5 L |
£132.08
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15672850
|
Honeywell-Chromasolv
270547-4X2.5L |
4 x 2.5 L |
£442.66
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- For HPLC, spectrophotometry, and environmental analysis
- Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
- Low UV impurity profile
- Low impurity gradient LC baseline
- Very low levels of non-volatile impurities
Chemical Identifiers
68-12-2 | |
73.10 | |
ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
6228 | |
N,N-dimethylformamide |
C3H7NO | |
MFCD00003284 | |
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa | |
CHEBI:17741 | |
CN(C)C=O |
Specifications
-61°C | |
0.944g/mL | |
n20/D 1.430 | |
HCON(CH3)2 | |
2.7mmHg (20 °C) | |
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa | |
CN(C)C=O | |
73.10 | |
CHEBI:17741 | |
≥99.9% | |
N,N-Dimethylformamide, for HPLC |
68-12-2 | |
153°C | |
C3H7NO | |
MFCD00003284 | |
605365 | |
ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
N,N-dimethylformamide | |
6228 | |
73.09g/mol | |
glass distillation |
Safety and Handling
P201-P280-P305 + P351 + P338-P308 + P313
H226-H312 + H332-H319-H360D
missing translation for 'einecsNumber' : 200-679-5
missing translation for 'rtecsNumber' : LQ2100000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- N,N-Dimethylformamide
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
Hazard Statement
- H226-Flammable liquid and vapour.
- H301-Toxic if swallowed.
- H319-Causes serious eye irritation.
- H360D-May damage the unborn child.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : N,N-dimethylformamide