Learn More
N-N-Butyldiethanolamine, 98%, Thermo Scientific Chemicals
27.24 EUR - 131.66 EUR
Chemical Identifiers
CAS | 102-79-4 |
---|---|
Molecular Formula | C8H19NO2 |
Molecular Weight (g/mol) | 161.24 |
MDL Number | MFCD00002856 |
InChI Key | GVNHOISKXMSMPX-UHFFFAOYSA-N |
Synonym | n-butyldiethanolamine, butyldiethanolamine, ethanol, 2,2'-butylimino bis, bide, 2,2'-butylimino diethanol, n,n-bis 2-hydroxyethyl butylamine, butylbis 2-hydroxyethyl amine, n-butyl-2,2'-iminodiethanol, 2,2-butylimino diethanol, n-butyl-n,n-bis hydroxyethyl amine |
PubChem CID | 7620 |
IUPAC Name | 2-[butyl(2-hydroxyethyl)amino]ethanol |
SMILES | CCCCN(CCO)CCO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10031740
|
Thermo Scientific Acros
154362500 |
250 mL | Glass bottle |
27.24 EUR
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10276100
|
Thermo Scientific Acros
154360010 |
1 L | Glass bottle |
66.93 EUR
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10237023
|
Thermo Scientific Acros
154360025 |
2.5 L | Glass bottle |
131.66 EUR
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
102-79-4 | |
161.24 | |
GVNHOISKXMSMPX-UHFFFAOYSA-N | |
7620 | |
CCCCN(CCO)CCO |
C8H19NO2 | |
MFCD00002856 | |
n-butyldiethanolamine, butyldiethanolamine, ethanol, 2,2'-butylimino bis, bide, 2,2'-butylimino diethanol, n,n-bis 2-hydroxyethyl butylamine, butylbis 2-hydroxyethyl amine, n-butyl-2,2'-iminodiethanol, 2,2-butylimino diethanol, n-butyl-n,n-bis hydroxyethyl amine | |
2-[butyl(2-hydroxyethyl)amino]ethanol |
Specifications
102-79-4 | |
100.0 | |
Brown to Brown-Red | |
264.0°C to 278.0°C | |
Authentic | |
C8H19NO2 | |
1.4615 to 1.4635 | |
04, 285 | |
n-butyldiethanolamine, butyldiethanolamine, ethanol, 2,2'-butylimino bis, bide, 2,2'-butylimino diethanol, n,n-bis 2-hydroxyethyl butylamine, butylbis 2-hydroxyethyl amine, n-butyl-2,2'-iminodiethanol, 2,2-butylimino diethanol, n-butyl-n,n-bis hydroxyethyl amine | |
GVNHOISKXMSMPX-UHFFFAOYSA-N | |
2-[butyl(2-hydroxyethyl)amino]ethanol | |
7620 | |
98% | |
N-N-Butyldiethanolamine |
97.5 | |
-70.0°C | |
0.9680g/mL | |
143°C | |
97.5% min. (GC) | |
CH3(CH2)3N(CH2CH2OH)2 | |
MFCD00002856 | |
0.968 | |
Solubility in water: miscible | |
CCCCN(CCO)CCO | |
161.24 | |
161.24 | |
Powder |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-055-
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only