missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Heptane, 99%, Optima™ for HPLC and GC, Fisher Chemical™
61.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
124.56 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
54.76 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Use promo code "22914" to get your promotional price.
124.56 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
54.76 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Click to view available options
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Description
- Screened for fluorescing contaminants, and UV-absorbing contaminants in the 197-300nm range, to reduce interferences for smoother baselines
- Tested by GC-ECD to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis
- Filtered through a sub-micron filter to remove particulates
- Intended for research use only
Specifications
Melting Point | -91°C |
Color | Colorless |
Boiling Point | 98°C |
Quantity | 2.5 L |
Packaging | Amber Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 48 mbar at 20°C |
Viscosity | 0.4 mPaS at 20°C |
Formula Weight | 100.2g/mol |
Safety and Handling
Product Identifier
- Heptane
- Danger
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P273-Avoid release to the environment.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'