missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ubinuclein 1, Mouse, Clone: UBN1-02, Abnova™
Mouse monoclonal antibody raised against partial recombinant Ubinuclein 1.
Brand: Abnova MAB6459.100ug
Additional Details : Weight : 0.02000kg
Specifications
Ubinuclein 1 | |
Monoclonal | |
Unconjugated | |
Immunoprecipitation Western Blot (1-5 ug/mL) The optimal working dilution should be determined by the end user. | |
Mouse | |
100 μg | |
Primary | |
Store at 4°C. Do not freeze. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunoprecipitation, Western Blot | |
UBN1-02 | |
Mouse monoclonal antibody raised against partial recombinant Ubinuclein 1. | |
In PBS, pH 7.4 (0.09% sodium azide) | |
Recombinant protein corresponding to amino acids 1-190 of Ubinuclein 1. | |
RUO | |
Human | |
Liquid |