missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MORC3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24994-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein that localizes to the nuclear matrix. The protein also has RNA binding activity, and has a predicted coiled coil domain.Specifications
MORC3 | |
Polyclonal | |
Unconjugated | |
MORC3 | |
KIAA0136, MORC3, NXP2, ZCW5, ZCWCC3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23515 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q14149 | |
MORC3 | |
MORC3 Fusion Protein Ag21825 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |