Learn More
Micronova™ Mop LoPro controlled environment NovaPoly 406mm MegaSwat
The MegaSwat™ mop is ideal for cleaning around heavy process equipment or calibrate apparatus. Features a flexible polytheylene interior with a choice of slipcovers
Brand: Micronova™ MS6-16NS
This item is not returnable.
View return policy
Description
- NovaPoly cover for use with MegaSwat™ mop
- Ideal for cleaning around heavy process equipment or calibrated apparatus
- 100% smooth knit polyester
- Compatible with strong disinfectants
- Autoclavable
Specifications
Non-sterile |