missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(V) isopropoxide, 99+% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7051540
Click to view available options
Quantity:
0.5 g
2 g
10 g
This item is not returnable. View return policy
209733-38-0
C15H35MoO5
391.39
MFCD00210636
HLFWWCCBFJUYJL-UHFFFAOYSA-N
molybdenum v isopropoxide, acmc-20akdz, pentaisopropoxymolybdenum v, molybdenum 5+ pentakis propan-2-olate
18475294
molybdenum;propan-2-olate
CC(C)O[Mo](OC(C)C)(OC(C)C)(OC(C)C)OC(C)C
This item is not returnable. View return policy
209733-38-0
C15H35MoO5
391.39
MFCD00210636
HLFWWCCBFJUYJL-UHFFFAOYSA-N
molybdenum v isopropoxide, acmc-20akdz, pentaisopropoxymolybdenum v, molybdenum 5+ pentakis propan-2-olate
18475294
molybdenum;propan-2-olate
CC(C)O[Mo](OC(C)C)(OC(C)C)(OC(C)C)OC(C)C

Precursor used in sol-gel process

Molybdenum(V) isopropoxide, it is used as a Precursor used in sol-gel process. It is used in nanomaterials and thin films. It is used as a solution based product for printed electronics. It acts a catalyst for selective isobutane oxidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Black to Yellow
Linear Formula Mo[OCH(CH3)2]5
Quantity 0.5 g
UN Number UN3181
Sensitivity Moisture sensitive
Solubility Information It is soluble in hydrocarbons,reacts with alcohols,ketones and esters. It reacts exothermally with water.
Formula Weight 391.04
Percent Purity ≥99%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Molybdenum(V) isopropoxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'