Learn More
Miele Professional™ Mobile Injector, Module A 300/1
Module A 300/1 for the optimum loading of laboratory glassware
Brand: Miele Professional™ 11116900
Code : NEW
Additional Details : Weight : 5.70000kg
Description
- Suitable for laboratory glassware such as Erlenmeyer flasks
- equipped with 8 injector nozzles with plastic supports ID 220
- Nozzle spacing 120 mm across the width, 123 mm across the depth