Learn More
Merck Millipore Milliflex™ Funnel without Membrane
Eliminates time-consuming washing, bagging, autoclaving and equipment handling. Milliflex™ Funnels without Membranes combine broad chemical compatibility with low protein binding.
Brand:  Merck Millipore MSP010022
Additional Details : Weight : 0.70880kg
Product Code. 12851756
  • £171.00 / Pack of 24
In stock, ready to ship: 3
Description
Description
  • Pre-sterilized and ready to use
  • Applications include clinical labs, food and beverage industry, industrial QC, environmental analysis, bioburden testing and water monitoring
Specifications
Specifications
Plastic
Milliflex Funnel w/o Membrane
Clinical Lab, Food and Beverage, Industrial QC, Environmental Analysis, Laboratory Filtration, Bioburden Testing, Water Monitoring
24/Pk.