missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ FE20 FiveEasy™ Benchtop pH Meter

Used for pH measurement in agriculture, food, beverage and water applications.  Mettler Toledo™ FE20 FiveEasy™ Benchtop pH Meter is a compact instrument that is versatile and easy to operate.

£334.40 - £458.70

Specifications

Accuracy (pH) ±0.01
Calibration Points 1 or 2
Display Type LCD
Range (pH) 0 to 14
Resolution (pH) 0.01
View More Specs

Includes: Meter Only package comes with meter, operating instructions, Quick Guide, power adapter.
Meter Kit package includes meter, operating instructions, Quick Guide, power adapter, LE409 pH electrode (01-915-676), electrode arm, 2 each pH 4.01 and pH 7.00 buffer sachets
ATC Kit package includes meter, operating instructions, Quick Guide, power adapter, LE438 pH electrode (01-911-250), electrode arm, 2 each pH 4.01 and pH 7.00 buffer sachets

Products 3
Product Code Brand Includes Range (mV) Type Price Quantity & Availability  
Product Code Brand Includes Range (mV) Type Price Quantity & Availability  
10385343
View accessory products View Documents
Mettler Toledo™
51302910
Meter, operating instructions, quick guide 0 to 1999 mV FE20 ph/mV/°C Meter Only This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10755384
View Documents
Mettler Toledo™
51302911
Meter, operating instructions, quick Guide, power adapter, LE409 pH electrode (01-915-676), electrode arm, 2 each pH 4.01 and pH 7.00 buffer sachets 0 to 1999 mV FE20 ph/mV/°C Meter Kit This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10526655
View Documents
Mettler Toledo™
51302916
Meter, operating instructions, Quick Guide, power adapter, LE438 pH electrode (01-911-250), electrode arm, 2 each pH 4.01 and pH 7.00 buffer sachets -1999 to 1999 mV FE20 pH ATC Kit This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Powerful, time-saving features

  • Automatic or manual termination of a measurement or calibration
  • Automatic calibration with predefined pH buffer groups
  • Automatic or manual temperature compensation
  • Two-point calibration with automatic buffer recognition
Large and informative LCD
  • Simultaneous display of measurement, temperature and endpoint
  • Electrode icon provides information on electrode condition
  • Calibration/measurement icon shows when instrument is calibrating or carrying out measurements
  • Setup icon lets you enter and modify settings
Easy operation
  • Simple start and finish of measurements and calibrations
  • Fast changeover between pH and mV parameters
  • Intuitive adjustment of settings such as manual temperature compensation
  • Save and access measurements and calibration data at the push of a button
  • Separate on/off switch
  • Measures pH, mV and temperature
  • Laminated Quick Guide outlines operation at a glance

Variety of inputs and outputs

  • BNC input for combination or half-cell electrode
  • Reference input for separate reference half cell
  • Accepts redox electrode for oxidation-reduction measurements
  • ATC input for NTC 30kω probe
Durable housing, universal power supply
  • Rugged ABS housing
  • PC-reinforced window
  • Tough membrane keypad
  • 100 to 240V 50/60Hz power supply provides 9VDC to instrument

Specifications

Specifications

±0.01
LCD
0.01
BNC for pH combination or half cell, 2mm banana socket for separate reference half cell, Cinch/RCA jack for ATC probe (NTC 30kilohm)
ABS/PC Reinforced Window, Membrane Keypad
NTC
0.1°C
0°C to 100°C
1.3 lb.
1 or 2
0 to 14
2.1 in.
20 cm
2mm Banana socket for analog output
1mV
-32°F to +212°F
120/240 V
SDS
Product Certifications