missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ EasyPlus Easy pH Titrator

Used to automate and speed up simple acid/base titrations. The EasyPlus Easy pH Titrator is ideal for the determining the acidity content of beverages, alkalinity of cleaning reagents and chemicals, free fatty acid content of cooking oils, and alkalinity (m-value) and acidity (p-value) of water.

Brand:  METTLER TOLEDO™ 30060041

Product Code. 16279481

  • 5295.00 € / Each
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket

Includes: Standard delivery components:
  • EG11-BNC sensor pH aqueous
  • EasyPlus burette 20mL
  • EasyPlus tubing set instrument
  • EasyStir GT
  • EasyPlus titration head GT
  • EasyPlus drying tube
  • Valve
  • Bottle head
  • Insert set titration head GT
  • Burette cover set
This item is not returnable. View return policy

Description

Description

  • Swiss design and construction in accordance with highest software and hardware standards
  • High quality component construction, capable of enduring harsh laboratory conditions
  • Small design capable of performing any basic titration with ease
  • Built-in iTitrate™ intelligence requires few parameters to be set before the instrument is ready for titration
  • Gives accurate, reproducible and repeatable results
  • Shows results automatically which are stored and are available for print or transfer to EasyDirect™ software
  • Smartphone-style Apps design is used to control all major instrument functions making it easy to use
  • LongClick™ helps to start the last analysis run without always navigating the routine analyses
  • EasySetup guides steps for installation and programming
  • Simple and intuitive navigation speaks 14 different languages including Arabic, Chinese, Thai, Portuguese and English
  • Reduces training time with the help of App style user interface while increasing work speed and simplicity
  • Meets all acid/base titration needs
TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.1
0.01
Yes
10 mL, 20 mL (Standard Delivery), or 50 mL
4.10/9.21/11.00; 1.68/4.00/10.01; 1.680/4.003/12.460; or User defined
BNC, USB A, USB B, Mini-DIN, RCA
Export via USB, USB Printer
4.3 inch, 480 x 272 pixel RGB
Detection of up to 3
EG11-BNC sensor pH aqueous, EasyPlus burette 20mL, EasyPlus tubing set instrument, EasyStir GT, EasyPlus titration head GT, EasyPlus drying tube, valve, bottle head, insert set titration head GT, burette cover set
Touchscreen
PP GF30
Easy pH
0.5% RSD
0 to 14
±0.1mV
0.1°C
User Defined
41 to 104°F
5 to 40°C
Yes
iTitrate
Yes
12 months
6.7 in.
8.66 in.
100 to 240V ±10% 50/60Hz
50/60 Hz
EasyPlus burette 20 mL (included)
0.2°C
Variable or fixed value
2
CE, CSA
Maximum number of samples in a series is 5. When choosing this option the mean value and RSD is calculated.
Easy pH
EasyPlus Automated Titrator
Potentiometric - acid/base titrations
BNC, USB A, USB B, Mini-DIN, RCA
15
Acid/base endpoint or inflection/equivalence point titrations
24 VDC / 1.25 A
±2000 mV
Max. 80% at 31°C
±0.1mV
None, Fixed, Variable
Low, Medium, High, Maximum
0 to 100°C
°C / °F
Normal, Fast, Cautious, Very cautious, User defined
Direct, back compensated, blank compensated
100 mL Beaker
220 x 170 x 350 mm
170 mm
220 mm
100 to 240V ±10%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers