Learn More
Thermo Scientific™ MEPS™ for LC: Thermo Scientific, CTC Analytics, HTA and Varian 8400 Systems
Save hours in sample preparation, and go from extraction to injection in a single process with these 250µL syringes for LC applications using CTC Analytics.
£245.95 - £262.00
Specifications
For Use With (Equipment) Thermo Scientific, CTC Analytics, HTA, and Varian 8400 Systems Using 100 µL and 250 µL MEPS Syringes
Type MEPS Barrel Insert and Needle (BIN)
View More Specs
Products 9
Product Code Brand Sorbent Description Price Quantity & Availability  
Product Code Brand Sorbent Description Price Quantity & Availability  
10015724
View Documents Promotion Details
Thermo Scientific™
60308-401
HyperSep Retain PEP MEPS Retain PEP MEPS for LC This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10299164
View Documents Promotion Details
Thermo Scientific™
60308-402
HyperSep Retain CX MEPS Retain-CX MEPS for LC This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10774817
View Documents Promotion Details
Thermo Scientific™
60308-403
HyperSep Retain AX MEPS Retain-AX MEPS for LC This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10649665
View Documents Promotion Details
Thermo Scientific™
60308-404
HyperSep Hypercarb MEPS Hypercarb MEPS for LC This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10784817
View Documents Promotion Details
Thermo Scientific™
60308-405
HyperSep Verify CX MEPS Verify-CX MEPS for LC This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10698895
View Documents Promotion Details
Thermo Scientific™
60308-406
HyperSep Verify AX MEPS Verify-AX MEPS for LC This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10219224
View Documents Promotion Details
Thermo Scientific™
60308-407
HyperSep C18 MEPS C18 MEPS for LC This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10449624
View Documents Promotion Details
Thermo Scientific™
60308-408
HyperSep Silica MEPS Silica MEPS for LC This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11894293
View Documents Promotion Details
Thermo Scientific™
60308-409
HyperSep Retain PEP, Retain CX, Retain AX, Hypercarb, and C18 Development kit; Retain PEP,-CX,-AX,Hypercarb, C18 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
Save hours in sample preparation, and go from extraction to injection in a single process with Thermo Scientific™ HyperSep™ MEPS™ (Micro Extraction by Packed Sorbent) technology.
Suitable for reversed phases, normal phases, mixed mode or ion exchange chemistries, Thermo Scientific HyperSep MEPS Syringes and Components for Analytics are available in a variety of SPE phase chemistries. The HyperSep MEPS Barrel Insert and Needle Assembly (BIN) contains the stationary phase, and is built into the syringe needle.
These HyperSep MEPS BINs are designed for use with Thermo Scientific, CTC Analytics, HTA and Varian 8400 LC Systems.
MEPS performs the same functions as SPE — the removal of interfering matrix components and the selective isolation and concentration of analytes — except that it miniaturizes conventional SPE packed bed devices from milliliter bed volumes to microliter volumes, and incorporates them right into the syringe.
Mechanically robust, the technique offers significant advantages over conventional SPE extraction for method development and other GC applications:
  • Significantly reduces the time to prepare and inject samples from hours to minutes — typically less than two minutes per sample
  • Eliminates all extra steps between sample preparation and sample injection; the technique is fully compatible with direct injection onto GC
  • Reduces buffer and solvent volume from milliliters to microliters
  • Reduces the required sample volume to as little as 3.6µL, making the techniques especially well-suited for on-line use with volume-limited samples
  • Can easily be automated compatible and is compatible with CTC and eVol platforms
Specifications
Specifications
Thermo Scientific, CTC Analytics, HTA, and Varian 8400 Systems Using 100 µL and 250 µL MEPS Syringes
MEPS Barrel Insert and Needle (BIN)