missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MELK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11403-1-AP-150UL
This item is not returnable.
View return policy
Description
The final step in heme metabolism in mammals is catalyzed by the cytosolic biliverdin reductase enzymes A and B (EC 1.3.1.24).Specifications
MELK | |
Polyclonal | |
Unconjugated | |
MELK | |
hMELK, HPK38, KIAA0175, MELK, Protein kinase PK38 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17279, 362510, 9833 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q14680, Q61846 | |
Melk | |
MELK Fusion Protein Ag1897 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |