missing translation for 'onlineSavingsMsg'
Learn More

MCM3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen MCM3
Concentration 0.18 mg/mL
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16894834
View Documents
Proteintech
15597-1-AP-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16884834
View Documents
Proteintech
15597-1-AP-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The protein encoded by this gene is one of the highly conserved mini-chromosome maintece proteins (MCM) that are involved in the initiation of eukaryotic genome replication. The hexameric protein complex formed by MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. This protein is a subunit of the protein complex that consists of MCM2-7. It has been shown to interact directly with MCM5/CDC46. This protein also interacts with, and thus is acetlyated by MCM3AP, a chromatin-associated acetyltransferase. The acetylation of this protein inhibits the initiation of DNA replication and cell cycle progression.
Specifications

Specifications

MCM3
Western Blot, Immunoprecipitation
Unconjugated
Rabbit
Rat, Human, Mouse
P25205, P25206
17215, 316273, 4172
MCM3 Fusion Protein Ag7966
Primary
-20°C
MCM3
0.18 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
HCC5, MCM3, P1 MCM3, P1.h, p102, RLF subunit beta, RLFB
Mcm3
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications