missing translation for 'onlineSavingsMsg'
Learn More

MAVS VISA Mouse anti-Human, Unconjugated, Clone: 1A8E9, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen MAVS VISA
Clone 1A8E9
Concentration 900 μg/mL
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16809205
View Documents
Proteintech
66911-1-IG-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16899195
View Documents
Proteintech
66911-1-IG-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The MAVS; VISA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MAVS; VISA. This antibody recognizes human antigen. The MAVS; VISA antibody has been validated for the following applications: IHC, WB, ELISA analysis.

Specifications

Specifications

MAVS VISA
900 μg/mL
Monoclonal
Mouse
57506
Primary
1A8E9
Western Blot, Immunohistochemistry, Immunofluorescence
Unconjugated
Human
IgG1
Videos
SDS
Documents

Documents

Product Certifications