Learn More
MAVS VISA Mouse anti-Human, Unconjugated, Clone: 1A8E9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66911-1-IG-150UL
This item is not returnable.
View return policy
Description
The MAVS; VISA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MAVS; VISA. This antibody recognizes human antigen. The MAVS; VISA antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
MAVS VISA | |
Monoclonal | |
900 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, Immunofluorescence | |
1A8E9 | |
Unconjugated | |
150 μL | |
57506 | |
IgG1 |