Learn More
Eppendorf™ Mastercycler™ PRO
£4505.00
Products 1
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
11864001
View Documents
Eppendorf™
6321000515
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
Reproducibility, specificity and speed - are the requirements for PCR in any application. The Mastercycler pro is unparalleled in its ability to fulfill these requirements. The proprietary vapo. protect™ technology reduces evaporation to a minimum. Concentrations in your PCR master mix remain consistent leading to stable and reproducible specificity. Non-specific binding is minimised beyond importance. The extremely high heating and cooling rates of the Mastercycler pro S give you the speed you need. Unsurpassed speed can be achieved with Eppendorf's Impulse PCR, a device driven hot-start function that increases heating rates to 8°C/s.
  • Ultimate reduction of evaporation
  • Extremely fast heating and cooling rates
  • Gradient blocks with SteadySlope technology
  • Intuitive graphic programming
  • Display to indicate cycler number in a network
  • Upgradable to real-time PCR*
  • Optional self-test of peltier elements
SDS