missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One MASTERBLOCK™ 96 Deep Well Conical Bottom 0.5mL Storage Plate

Suitable for cultivating bacteria or yeasts. Greiner Bio-One MASTERBLOCK™ 96 Deep Well Conical Bottom 0.5mL Storage Plate is also suitable for storing sample material.

£633.00

Specifications

Format Standard
Array 8 x 12
Color Natural
Material Polypropylene
No. of Wells 96
View More Specs

Products 1
Product Code Brand Sterility Price Quantity & Availability  
Product Code Brand Sterility Price Quantity & Availability  
10650973
View Documents
Greiner Bio-One
786261
Sterile
£633.00
Pack of 80
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Sealable with adhesive films and heat sealer
  • High temperature resistance
  • High chemical resistance
  • A
Specifications

Specifications

Standard
Natural
96
Conical Bottom (V-well)
80/Cs.
8 x 12
Polypropylene
0.5 mL
80
Videos
SDS
Documents

Documents

Product Certifications