missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Maspin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
136.48 CHF - 385.92 CHF
Spezifikation
Antigen | Maspin |
---|---|
Konzentration | 0.15 mg/mL |
Anwendungen | Immunohistochemistry (Paraffin), Western Blot |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16898983
|
Proteintech
11722-1-AP-20UL |
20 μL |
136.48 CHF
20 Mikroliter |
Lager vorrätigLoggen Sie sich ein, um den Lagerbestand zu sehen
|
|||||
16888983
|
Proteintech
11722-1-AP-150UL |
150 μL |
385.92 CHF
150 Mikroliter |
Verfügbar ab: 16-12-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
SERPINB5 is a tumor suppressor. It blocks the growth, invasion, and metastatic properties of mammary tumors. As it does not undergo the S (stressed) to R (relaxed) conformational transition characteristic of active serpins, it exhibits no serine protease inhibitory activity.Spezifikation
Maspin | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P36952 | |
5268 | |
Maspin Fusion Protein Ag2304 | |
Primary | |
-20°C | |
SERPINB5 |
0.15 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
maspin, Peptidase inhibitor 5, PI 5, PI5, Serpin B5, SERPINB5, SPB5 | |
SERPINB5 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |