missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Maspin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11722-1-AP-20UL
This item is not returnable.
View return policy
Description
SERPINB5 is a tumor suppressor. It blocks the growth, invasion, and metastatic properties of mammary tumors. As it does not undergo the S (stressed) to R (relaxed) conformational transition characteristic of active serpins, it exhibits no serine protease inhibitory activity.Specifications
Maspin | |
Polyclonal | |
Unconjugated | |
SERPINB5 | |
maspin, Peptidase inhibitor 5, PI 5, PI5, Serpin B5, SERPINB5, SPB5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5268 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P36952 | |
SERPINB5 | |
Maspin Fusion Protein Ag2304 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |