missing translation for 'onlineSavingsMsg'
Learn More

Magnesium chloride hexahydrate, BAKER ANALYZED™ ACS, J.T. Baker™

Meets ACS Specifications

£71.80 - £111.00

Chemical Identifiers

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10785921
View Documents
JT Baker
0162.0250
250 g
£71.80
250g
Please to purchase this item. Need a web account? Register with us today!
10795921
View Documents
JT Baker
0162.1000
1 kg
£111.00
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

7791-18-6
203.30
DHRRIBDTHFBPNG-UHFFFAOYSA-L
O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]
Cl2H12MgO6
MFCD00149781
magnesium(2+) hexahydrate dichloride

Specifications

1412°C (1013 hPa)
7791-18-6
Cl2H12MgO6
DHRRIBDTHFBPNG-UHFFFAOYSA-L
magnesium(2+) hexahydrate dichloride
1.57 g/cm³ (25°C)
117°C
99.0 to 102.0%
MFCD00149781
O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]
203.30
Videos
SDS
Documents

Documents

Product Certifications