Learn More
Lovibond™ MC500, MD600 Colorimeter Replacement Reagents: pH value
For use with MC500 and MD600 multi-parameter colorimeter
Brand: Lovibond™ 471040
Code : NEW
Additional Details : Weight : 0.50000kg
Specifications
Red | |
15 mL | |
Water quality testing | |
6.5 to 8.4 | |
Phenol red | |
0.5 oz. |
7.7 | |
Liquid | |
MC500 multi-parameter colorimeter | |
20°C | |
Phenol Red Solution |
Safety and Handling
- Phenolred solution
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P233-Keep container tightly closed.
Supplemental information
- MIXTURE LIST-Contains : butan-2-ol