missing translation for 'onlineSavingsMsg'
Learn More

BioTek™ Lionheart™ FX Automated Microscope

Lionheart FX Automated Live Cell Imager is an all-inclusive automated digital microscope for live cell biologists, capturing images and transforming them into intelligent information.

Brand:  BioTek™ LFX


Additional Details : Weight : 26.30000kg

Product Code. 15788469

  • £55645.00 / Each

Estimated Shipment: 02-09-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

With Lionheart FX, you can capture, analyze, annotate and produce videos with ease. This all-inclusive microscopy system is optimized for live cell imaging with up to 100x air and oil immersion magnification. Brightfield, color brightfield, phase contrast and fluorescence channels offer maximum support for a wide range of imaging applications.
  • Automated digital microscopy with up to 100x magnification 
  • Brightfield, color brightfield, phase contrast and fluorescence with image and laser autofocus
  • Powerful Gen5 3.0 software with seamless image capture, annotation, analysis and movie maker, requiring no extensive training
  • Kinetic live cell assay support with temperature and CO2/O2 control, humidity and reagent injectors
  • Fully integrated and compact design offers quick installation and setup
  • Able to read 6- to 1536-well micro plates
  • Supports Microscope slides, Petri and cell culture dishes, cell culture flasks (T25, T75), counting chambers (hemocytometers), chamber slides, and other labware up to 1.5 in. tall

Certifications

CE and TUV

Compliance

RoHS

Specifications

Specifications

16-bit gray scale, Sony CCD, 1.25 megapixel
High power LEDs (available wavelengths: 365nm, 390nm, 465nm, 505nm, 523nm, 590nm, 623nm, 655nm, 740nm)
18.3 in.
Live cell imaging
35.8 cm
Gen5 Microplate Reader and Imager
45.5 cm
Cell Imager
16-bit gray scale, Sony CCD, 1.25 megapixel
46.5 cm
14.4 in.
High power LEDs (available wavelengths: 365nm, 390nm, 465nm, 505nm, 523nm, 590nm, 623nm, 655nm, 740nm)
17.9 in.
Videos
SDS
Documents
Certificates