missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIMK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19699-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody has no cross-reaction with LIMK2.
There are approximately 40 known eukaryotic LIM proteins, so named for the LIM domains they contain. LIM domains are highly conserved cysteine-rich structures containing 2 zinc fingers. Although zinc fingers usually function by binding to DNA or RNA, the LIM motif probably mediates protein-protein interactions. LIM kinase-1 and LIM kinase-2 belong to a small subfamily with a unique combination of 2 N-terminal LIM motifs and a C-terminal protein kinase domain. LIMK1 is likely to be a component of an intracellular signaling pathway and may be involved in brain development. LIMK1 hemizygosity is implicated in the impaired visuospatial constructive cognition of Williams syndrome.Specifications
LIMK1 | |
Polyclonal | |
Unconjugated | |
LIMK1 | |
LIM domain kinase 1, LIMK, LIMK 1, LIMK1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16885, 3984, 65172 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P53667, P53668, P53669 | |
LIMK1 | |
Peptide | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |