Learn More
LIMK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 19699-1-AP-150UL
 View more versions of this product
Product Code. 16889174
  • £349.00 / 150µL
Explore more special offers
Description
Description
The antibody has no cross-reaction with LIMK2.
There are approximately 40 known eukaryotic LIM proteins, so named for the LIM domains they contain. LIM domains are highly conserved cysteine-rich structures containing 2 zinc fingers. Although zinc fingers usually function by binding to DNA or RNA, the LIM motif probably mediates protein-protein interactions. LIM kinase-1 and LIM kinase-2 belong to a small subfamily with a unique combination of 2 N-terminal LIM motifs and a C-terminal protein kinase domain. LIMK1 is likely to be a component of an intracellular signaling pathway and may be involved in brain development. LIMK1 hemizygosity is implicated in the impaired visuospatial constructive cognition of Williams syndrome.
Specifications
Specifications
LIMK1
Polyclonal
Unconjugated
LIMK1
LIM domain kinase 1, LIMK, LIMK 1, LIMK1
Rabbit
Antigen Affinity Chromatography
RUO
16885, 3984, 65172
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P53667, P53668, P53669
LIMK1
Peptide
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Ofertas especiales
Ofertas especiales