Learn More
Ansell™ Lightweight G01R Gloves
Click to view available options
Size: 7.5
Description
- Lightweight natural rubber
- Inside slightly flocked for very good touch
- Tear-resistant rolled edge for easy and quick donning
- Cuff with grooves as air ducts for excellent ventilation while protecting the forearm
- Hammer impact profile for very good wet and dry grip
- Ideal for use in the laboratory, for short-term contact with chemicals, for light maintenance, for painting and varnishing, in the printing industry, the pharmaceutical industry
Specifications
Specifications
Color | Red |
Material | Latex |
Size | 7.5 |
Quantity | 12 pairs |
Length (Metric) | 325 mm |