missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Lawesson's Reagent, 97%

Catalog Number p-7021973
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
19172-47-5
C14H14O2P2S4
404.452
MFCD00005171
CFHGBZLNZZVTAY-UHFFFAOYSA-N
lawesson's reagent, lawesson reagent, 2,4-bis 4-methoxyphenyl-1,3,2,4-dithiadiphosphetane 2,4-disulfide, unii-a4125mq8rx, 1,3,2,4-dithiadiphosphetane, 2,4-bis 4-methoxyphenyl-, 2,4-disulfide, 2,4-bis 4-methoxyphenyl-2,4-dithioxo-1,3,2,4-dithiadiphosphetane, 2,4-bis 4-methoxyphenyl-1,3,2,4-dithiadiphosphetane-2,4-disulfide, 2,4-bis 4-methoxyphenyl-1,3-dithia-2,4-diphosphetane 2,4-disulfide, 2,4-bis 4-methoxyphenyl-1,3-dithia-2,4-diphosphetane 2,4-disulphide, 4-methoxyphenylthiophosphoric cyclic di thioanhydride
87949
2,4-bis(4-methoxyphenyl)-2,4-bis(sulfanylidene)-1,3,2$l^{5},4$l^{5}-dithiadiphosphetane
COC1=CC=C(C=C1)P2(=S)SP(=S)(S2)C3=CC=C(C=C3)OC
19172-47-5
C14H14O2P2S4
404.452
MFCD00005171
CFHGBZLNZZVTAY-UHFFFAOYSA-N
lawesson's reagent, lawesson reagent, 2,4-bis 4-methoxyphenyl-1,3,2,4-dithiadiphosphetane 2,4-disulfide, unii-a4125mq8rx, 1,3,2,4-dithiadiphosphetane, 2,4-bis 4-methoxyphenyl-, 2,4-disulfide, 2,4-bis 4-methoxyphenyl-2,4-dithioxo-1,3,2,4-dithiadiphosphetane, 2,4-bis 4-methoxyphenyl-1,3,2,4-dithiadiphosphetane-2,4-disulfide, 2,4-bis 4-methoxyphenyl-1,3-dithia-2,4-diphosphetane 2,4-disulfide, 2,4-bis 4-methoxyphenyl-1,3-dithia-2,4-diphosphetane 2,4-disulphide, 4-methoxyphenylthiophosphoric cyclic di thioanhydride
87949
2,4-bis(4-methoxyphenyl)-2,4-bis(sulfanylidene)-1,3,2$l^{5},4$l^{5}-dithiadiphosphetane
COC1=CC=C(C=C1)P2(=S)SP(=S)(S2)C3=CC=C(C=C3)OC

Lawesson's reagent is a thiation agent used to convert carbonyl compounds into thiocarbonyls. It is also used to thionate enones, esters, lactones, amides, lactams and quinones. Further, it is used to prepare thiols from alcohols. It is associated with silver perchlorate and utilized as and oxophilic Lewis acid catalyst for Diels-Alder reaction of dienes with alfa, beta-unsaturated aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lawesson′s reagent is a thiation agent used to convert carbonyl compounds into thiocarbonyls. It is also used to thionate enones, esters, lactones, amides, lactams and quinones. Further, it is used to prepare thiols from alcohols. It is associated with silver perchlorate and utilized as and oxophilic Lewis acid catalyst for Diels-Alder reaction of dienes with alfa, beta-unsaturated aldehydes.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 221°C to 225°C
Quantity 5 g
UN Number UN3134
Beilstein 1024888
Sensitivity Moisture sensitive
Merck Index 14,5391
Solubility Information Insoluble in water.
Formula Weight 404.47
Percent Purity 97%
Chemical Name or Material Lawesson's Reagent

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'