missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Laminated Polyester Biohazard Label
Brand: Brady™ 227308
Code : Z4
Additional Details : Weight : 0.01000kg
Specifications
Black on Yellow | |
Triangular | |
Laminated Polyester | |
Permanent Acrylic | |
0.099 mm | |
-40 °C-110 °C |
Global | |
Gloss | |
250/Pk. | |
87 mm | |
100.00 mm |