missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD71/Transferrin Receptor Ab-3, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry applications with Epredia™ CD71/Transferrin Receptor Ab-3, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-1096-R7

 View more versions of this product

Product Code. 12648936

  • 554.00 € / 7mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Cell surface transferrin receptor (CD71) is a disulfide-bonded homodimeric glycoprotein. Ligand for transferrin receptor is the serum iron transport protein, transferrin. This receptor is broadly distributed in carcinomas, sarcomas, leukemias, and lymphomas. Transferrin receptor (CD71) has been reported to be associated with cell proliferation in both normal and neoplastic tissues and useful in predicting clinical behavior or response to therapy in a number of malignancies including breast cancer.

Host Species: Mouse

Clone: 10F11

Isotype: IgG2b

Species Reactivity: Human. Others not known.

Epitope: Not determined

Molecular Weight: ∽90-95 kDa

Positive Control: Placenta

Cellular Localization: Granular cytoplasmic and membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

CD71/Transferrin Receptor Ab-3
Monoclonal
Unconjugated
7 mL
Primary
Prediluted
Immunohistochemistry (Paraffin)
10F11
Mouse
RUO
Human
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions