missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Ornithine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog Number p-3758609
18.46 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
3184-13-2
C5H12N2O2·HCl
168.62
MFCD00064562
GGTYBZJRPHEQDG-WCCKRBBISA-N
l-ornithine hydrochloride, l-ornithine monohydrochloride, h-orn-oh.hcl, l-ornithine hcl, l +-ornithine hydrochloride, l-ornithine, monohydrochloride, s-2,5-diaminopentanoic acid hydrochloride, ornithine hydrochloride van, unii-hbk84k66xh, ornithine l monochlorohydrate
76654
(2S)-2,5-diaminopentanoic acid;hydrochloride
C(CC(C(=O)O)N)CN.Cl
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
3184-13-2
C5H12N2O2·HCl
168.62
MFCD00064562
GGTYBZJRPHEQDG-WCCKRBBISA-N
l-ornithine hydrochloride, l-ornithine monohydrochloride, h-orn-oh.hcl, l-ornithine hcl, l +-ornithine hydrochloride, l-ornithine, monohydrochloride, s-2,5-diaminopentanoic acid hydrochloride, ornithine hydrochloride van, unii-hbk84k66xh, ornithine l monochlorohydrate
76654
(2S)-2,5-diaminopentanoic acid;hydrochloride
C(CC(C(=O)O)N)CN.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 245.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Plastic bottle
Linear Formula H2N(CH2)3CH(NH2)CO2H·HCl
Quantity 25 g
Beilstein 04, 420
Merck Index 14, 6874
Specific Rotation Condition + 24.00 (20.00°C c=4,6N HCl)
Specific Rotation + 24.00
Solubility Information (5% in 1 N HCl) Clear colorless
Formula Weight 168.62
Percent Purity 99%
Physical Form Powder
Chemical Name or Material L(+)-Ornithine hydrochloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'