missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15612-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a BTB and kelch domain containing protein and belongs to the kelch repeat domain containing superfamily of proteins. The encoded protein functions as an adaptor protein that complexes with Cullin 3 and other proteins to form the Cullin 3-based E3 ubiquitin-protein ligase complex. This complex is necessary for proper chromosome segregation and completion of cytokinesis. Alternate splicing results in multiple transcript variants.Specifications
KLHL13 | |
Polyclonal | |
Unconjugated | |
KLHL13 | |
BKLHD2, kelch like 13 (Drosophila), Kelch like protein 13, KIAA1309, KLHL13 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
313445, 67455, 90293 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q80TF4, Q9P2N7 | |
KLHL13 | |
KLHL13 Fusion Protein Ag8003 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |