Learn More
Ecolab™ Klerwipe™ Neutral Detergent
Sterile low particulate mop wipes impregnated with neutral detergent
Brand:  Ecolab™ 3079260
Code : NEW
Additional Details : CAS Number : 69013-18-9 Weight : 6.14000kg
Product Code. 15306128
  • £1140.00 / Pack of 10
Estimated Shipment: 09-05-2024
to see stock.
Add to basket
Description
Description
  • The detergent is 0.2 micron filtered then filled and bagged in a Grade C cleanroom before being terminally sterilised by gamma irradiation
  • A sterile ready-to-use detergent mop wipe
  • In a peelable pack
  • For large surfaces
Specifications
Specifications
Mop Wipe
420 x 250 mm
10 x 20 Wipes