Learn More
Iron(III) sulfate pentahydrate, 97%, Thermo Scientific Chemicals
48.84€ - 641.19€
Chemical Identifiers
CAS | 142906-29-4 |
---|---|
Molecular Formula | Fe2O12S3·5H2O |
Molecular Weight (g/mol) | 489.96 |
MDL Number | MFCD00149714 |
InChI Key | YHGPYBQVSJBGHH-UHFFFAOYSA-H |
Synonym | unii-3hws7hf5xd, 3hws7hf5xd, iron iii sulfate pentahydrate, sulfuric acid, iron 3+ salt 3:2 , pentahydrate 9ci, acmc-20n1we, ferric sulfate pentahydrate, iron iii sulphate pentahydrate, 2fe.3so4.5h2o, sulfuric acid, iron 3+ salt 3:2 , pentahydrate |
PubChem CID | 23443659 |
IUPAC Name | iron(3+);trisulfate;pentahydrate |
SMILES | O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3].[Fe+3] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10306741
|
Thermo Scientific Acros
345230050 |
5 g | Glass Bottle |
48.84€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10522354
|
Thermo Scientific Acros
345231000 |
100 g | Plastic bottle |
56.58€
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10460863
|
Thermo Scientific Acros
345235000 |
500 g | Plastic bottle |
118.59€
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13418528
|
Thermo Scientific Acros
345230051 |
5 kg | Plastic Drum |
641.19€
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
142906-29-4 | |
489.96 | |
YHGPYBQVSJBGHH-UHFFFAOYSA-H | |
23443659 | |
O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3].[Fe+3] |
Fe2O12S3·5H2O | |
MFCD00149714 | |
unii-3hws7hf5xd, 3hws7hf5xd, iron iii sulfate pentahydrate, sulfuric acid, iron 3+ salt 3:2 , pentahydrate 9ci, acmc-20n1we, ferric sulfate pentahydrate, iron iii sulphate pentahydrate, 2fe.3so4.5h2o, sulfuric acid, iron 3+ salt 3:2 , pentahydrate | |
iron(3+);trisulfate;pentahydrate |
Specifications
142906-29-4 | |
Crystalline or Granular Powder | |
Fe2O12S3·5H2O | |
MFCD00149714 | |
Solubility in water: soluble | |
O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3].[Fe+3] | |
489.96 | |
489.96 | |
Iron(III) sulfate pentahydrate |
Beige-Yellow to White | |
97% | |
Fe2(SO4)3·5H2O | |
unii-3hws7hf5xd, 3hws7hf5xd, iron iii sulfate pentahydrate, sulfuric acid, iron 3+ salt 3:2 , pentahydrate 9ci, acmc-20n1we, ferric sulfate pentahydrate, iron iii sulphate pentahydrate, 2fe.3so4.5h2o, sulfuric acid, iron 3+ salt 3:2 , pentahydrate | |
YHGPYBQVSJBGHH-UHFFFAOYSA-H | |
iron(3+);trisulfate;pentahydrate | |
23443659 | |
97% |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con
GHS Signal Word: Danger
- Iron(III)sulfate pentahydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H302-Harmful if swallowed.
- H290-May be corrosive to metals.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P362-Take off contaminated clothing.
RUO – Research Use Only