Learn More
Sigma Laborzentrifugen™ IQ/OQ Documentation for a Centrifuge
A comprehensive installation qualification for a tempered centrifuge with one rotor.
Brand: Sigma Laborzentrifugen™ 17711
This item is not returnable.
View return policy
Description
- Installation qualification (IQ) and operation qualification (OQ) are performed during production at the factory
- Includes the installation and verification of all of the stated parameters for one rotor
Specifications
Tempered centrifuge with one rotor | |
Installation/Operation Certificate |