missing translation for 'onlineSavingsMsg'
Learn More

Iodotrimethylsilane, 97%, stab. with copper, Thermo Scientific Chemicals

Product Code. 11485353
Click to view available options
Quantity:
5 g
25 g
100 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Iodotrimethylsilane is used for the introduction of trimethylsilyl group in organic synthesis. It is also useful for gas chromatography analysis by converting alcohol into a silyl ether derivative, thereby making it more volatile than the original molecule.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodotrimethylsilane is used for the introduction of trimethylsilyl group in organic synthesis. It is also useful for gas chromatography analysis by converting alcohol into a silyl ether derivative, thereby making it more volatile than the original molecule.

Solubility
Miscible with water.

Notes
Moisture and light Sensitive. Incompatible withstrong acids, strong bases, strong oxidizing agents and reacts violently with water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16029-98-4
Molecular Formula C3H9ISi
Molecular Weight (g/mol) 200.094
MDL Number MFCD00001028
InChI Key CSRZQMIRAZTJOY-UHFFFAOYSA-N
Synonym trimethylsilyl iodide, trimethyliodosilane, silane, iodotrimethyl, iodo trimethyl silane, trimethylsilyliodide, tms-iodide, iodotrimethysilane, iodotrimetylsilane, tms iodide, iodo trimethylsilane
PubChem CID 85247
IUPAC Name iodo(trimethyl)silane
SMILES C[Si](C)(C)I

Specifications

Boiling Point 105°C to 106°C
Quantity 250 g
Linear Formula (CH3)3SiI
UN Number UN2924
Beilstein 1731136
Flash Point −2°C (28°F)
Solubility Information Miscible with water.
Formula Weight 200.09
Percent Purity 97%
Refractive Index 1.476
Sensitivity Light and moisture sensitive
Density 1.44
Chemical Name or Material Iodotrimethylsilane, stabilized with copper
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'