Learn More
Cytiva Inlet filter holder kit
Use as filter holder for buffer inlet tubes. Cytiva Lifescience™ Inlet filter holder kit is an important chromatography accessory.
Brand: Cytiva 11-0004-07
Code : GX
Additional Details : Weight : 0.02160kg
Description
- The wetted parts are resistant to organic solvents and salt buffers commonly used in chromatography of biomolecules
- Except 100 % Ethyl acetate, 100 % Hexane and 100 % Tetrahydrofuran.
- Support screen Material Support net and filter: Polypropylene (PP)
- Filter holder: Titanium (Ti)