Learn More
IGHM Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11016-1-AP-150UL
 View more versions of this product
Product Code. 16877793
  • £349.00 / 150µL
Explore more special offers
Description
Description
This antibody detects the heavy chain of human IgM (∽75 kDa).
IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells.
Specifications
Specifications
IGHM
Polyclonal
Unconjugated
IGHM
IGHM, MU, VH
Rabbit
Antigen Affinity Chromatography
RUO
3507
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P01871
IGHM
IGHM Fusion Protein Ag1459
150 μL