missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Iron-Supplemented Newborn Calf Serum, New Zealand Origin
Contains three to four times as much available iron and transferrin as FBS or equine serum
£14.79 - £138.72
Specifications
Age | <10 days |
---|---|
Country of Origin | New Zealand |
Product Type | Newborn Calf Bovine Serum |
Sterility | Sterile |
Sterilization Method | Sequential Triple-filtered, 100 nm |
Product Code | Brand | Quantity | dimProcessing | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | dimProcessing | Price | Quantity & Availability | ||||
16655785
|
Cytiva
SH30626.01IR |
100 mL | N/A | ||||||
16665785
|
Cytiva
SH30626.02IR30-40 |
500 mL | N/A | ||||||
16675785
|
Cytiva
SH30626.03IR30-40 |
1000 mL | N/A | ||||||
11572830
|
Cytiva
SH30626.01 |
N/A | |||||||
Description
- Carefully collected, processed, and filtered in New Zealand to ensure safety against bovine diseases
- Contains three to four times as much available iron and transferrin as FBS or equine serums
- Collected from cattle that are typically less than 10 days old
Specifications
<10 days | |
Newborn Calf Bovine Serum | |
Sequential Triple-filtered, 100 nm | |
HyClone™ |
New Zealand | |
Sterile | |
Yes |