missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Fetal Bovine Serum, USDA Tested
Sourced from countries recognized as being free of BSE and FMD by the USDA
Brand: Cytiva SH30910.03
This item is not returnable.
View return policy
Description
Supplement cell culture growth with USDA-tested HyClone™ Fetal Bovine Serum (FBS), sourced from countries recognized as being free of BSE and FMD by the USDA.
Supplement cell culture growth with USDA-tested HyClone™ Fetal Bovine Serum (FBS), sourced from countries recognized as being free of BSE and FMD by the USDA.
Specifications
Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Inorganic Phosphorus, Lactate Dehydrogenase, Osmolality, pH, Potassium, SGOT, SGPT, Sodium, Total Bilirubin, Total Protein | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
United States | |
500 mL | |
Sequential-filtered, 100 nm | |
None | |
≤ 25 EU/mL | |
≤ 25 mg/dL | |
USDA Inspected |
USDA Regulations | |
USDA Tested Fetal Bovine Serum, Central American Sourced | |
Fetal Bovine Serum | |
Sterile | |
Characterized | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
No | |
No |