missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HuR Mouse anti-Human, Mouse, Rat, Clone: 1A11H2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66549-1-IG-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the ELAVL protein family. This encoded protein contains 3 RNA-binding domains and binds cis-acting AU-rich elements. It destabilizes mRNAs and thereby regulates gene expression.Specifications
HuR | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
B5DF91, P70372, Q15717 | |
Elavl1 | |
HuR Fusion Protein Ag2249 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG2a |
Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1A11H2 | |
Unconjugated | |
ELAVL1 | |
ELAV like protein 1, ELAV1, ELAVL1, Hu antigen R, Hua, HuR, MelG | |
Mouse | |
Protein A | |
RUO | |
15568, 1994, 363854 | |
-20°C | |
Liquid |