missing translation for 'onlineSavingsMsg'
Learn More

Heptane, 99%, Reagent Grade, Honeywell™

Product Code. 15607900
Click to view available options
Quantity:
500 mL
1 L
200 L
Packaging:
Glass Bottle
Metal Drum
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC

Specifications

Melting Point -90.5°C
Color Colorless
Density 0.684g/mL at 25°C
Boiling Point 98.2°C to 98.4°C (1000 hPa)
Quantity 1 L
Explosion Limit 0.07
Flash Point −4°C (24.8°F)
Ignition Point 433°F
Odor Characteristic
Packaging Glass Bottle
Physical Form Liquid
Linear Formula CH3(CH2)5CH3
Vapor Pressure 83mmHg (37.7°C), 40mmHg (20°C)
UN Number UN1206
Beilstein 1730763
Solubility Information 0.003g/L in water at 0°C
Formula Weight 100.2g/mol
Percent Purity 0.99
Chemical Name or Material Heptane
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'