Learn More
Kartell™ HDPE General Purpose Funnels
Manufactured from HDPE for optimum strength and chemical resistance. Kartell™ HDPE General Purpose Funnels provide safety and precision in filling bottles in the laboratory.
Brand:  Kartell™ 0017700
Additional Details : Weight : 1.87200kg
Product Code. 15733189
  • £114.00 / Pack of 10
Estimated Shipment: 14-05-2024
to see stock.
Description
Description
Manufactured from HDPE for optimum strength and chemical resistance. Kartell™ HDPE General Purpose Funnels provide safety and precision in filling bottles in the laboratory.
  • Suitable for a wide range of liquids
  • Upstand on rim reduces potential overflow
  • Outer ribbing eliminates back pressure
  • Hanging eye for storage