missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HBE1 Mouse anti-Human, FITC, Clone: 2C11G6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech FITC-66151-100UL
This item is not returnable.
View return policy
Description
This antibody specifically recognizes the HBE1 and doesn't cross-react with other globin chains. This antibody is conjugated with FITC.
The epsilon globin gene (HBE) is normally expressed in the embryonic yolk sac: two epsilon chains together with two zeta chains (an alpha-like globin) constitute the embryonic hemoglobin Hb Gower I; two epsilon chains together with two alpha chains form the embryonic Hb Gower II. Both of these embryonic hemoglobins are normally supplanted by fetal, and later, adult hemoglobin. The five beta-like globin genes are found within a 45 kb cluster on chromosome 11 in the following order: 5′-epsilon - G-gamma - A-gamma - delta - beta-3′Specifications
HBE1 | |
Monoclonal | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.3 | |
P02100 | |
HBE1 | |
Protein G | |
RUO | |
3046 | |
-20°C | |
Liquid |
ELISA | |
2C11G6 | |
FITC | |
HBE1 | |
HBE1, Hemoglobin epsilon, Hemoglobin epsilon chain | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |