Learn More
Fisherbrand™ Hard Surface Adhesive Mats
Ideal for maintaining a critical clean environment
£107.00
Specifications
Color Blue
Dimensions (L x W) 91 x 46 cm (36 x 18 in.)
View More Specs
Products 1
Product Code Brand Surface Coating Price Quantity & Availability  
Product Code Brand Surface Coating Price Quantity & Availability  
11952667
View Documents Promotion Details
Fisherbrand™
Moderate Tack
£107.00
Pack of 120
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
Description
Description
Ideal for maintaining a critical clean environment
  • Ideal for use in electronic, pharmaceutical, laboratory and food industry cleanrooms
  • When top sheet becomes soiled, peel it off to expose fresh, clean surface
  • Reorder notification on 18th sheet
  • Blue sheets on blue mat
  • Highly effective in dust removal and easy to use
  • Moderate Tack for standard clean needs (part number: 19-181-519)
  • High Tack for critical environment users (part number: 19-181-516)
  • Each mat has top and bottom cover sheets
Order Info
30 sheets per mat.
Specifications
Specifications
Blue
91 x 46 cm (36 x 18 in.)