Learn More
Brady™ HandiMark™ Indoor/Outdoor Vinyl Tape
£47.32 - £84.74
Specifications
Certifications/Compliance 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
Coating Powder-coated
Finish Gloss
For Use With (Application) For facility and safety identification
For Use With (Equipment) HandiMark Printer
View More Specs
Products 33
Product Code Brand Color Length (Metric) Width (Metric) Price Quantity & Availability  
Product Code Brand Color Length (Metric) Width (Metric) Price Quantity & Availability  
12830560
View Documents
Brady™
142287
Black 15 m 13 mm
£47.32
1 box
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Add to basket
12850570
View Documents
Brady™
142288
Black 15 m 25 mm
£67.06
1 box
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Add to basket
12870580
View Documents
Brady™
142289
Black 15 m 50 mm
£84.74
1 box
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Add to basket
12840560
View Documents
Brady™
142281
Blue 15 m 13 mm
£47.32
1 box
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Add to basket
12860570
View Documents
Brady™
142282
Blue 15 m 25 mm
£67.06
1 box
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Add to basket
12880580
View Documents
Brady™
142283
Blue 15 m 50 mm
£84.74
1 box
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Add to basket
12850560
View Documents
Brady™
142293
Brown 15 m 13 mm
£47.32
1 box
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Add to basket
12870570
View Documents
Brady™
142294
Brown 15 m 25 mm
£67.06
1 box
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Add to basket
12890580
View Documents
Brady™
142295
Brown 15 m 50 mm
£84.74
1 box
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Add to basket
12860560
View Documents
Brady™
142302
Clear 15 m 13 mm
£47.32
1 box
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Add to basket
12880570
View Documents
Brady™
142303
Clear 15 m 25 mm
£67.06
1 box
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Add to basket
12870560
View Documents
Brady™
142296
Gold 15 m 13 mm
£47.32
1 box
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Add to basket
12890570
View Documents
Brady™
142297
Gold 15 m 25 mm
£67.06
1 box
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Add to basket
12810590
View Documents
Brady™
142298
Gold 15 m 50 mm
£84.74
1 box
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Add to basket
12880560
View Documents
Brady™
142275
Green 15 m 13 mm
£47.32
1 box
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Add to basket
12800580
View Documents
Brady™
142276
Green 15 m 25 mm
£67.06
1 box
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Add to basket
12890560
View Documents
Brady™
142290
Gray 15 m 13 mm
£47.32
1 box
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Add to basket
12820590
View Documents
Brady™
142277
Green 15 m 50 mm
£84.74
1 box
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Add to basket
12810580
View Documents
Brady™
142291
Gray 15 m 25 mm
£67.06
1 box
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Add to basket
10795305
View Documents
Brady™
142292
Gray 15 m 50 mm
£84.74
1 box
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Add to basket
12840580
View Documents
Brady™
142306
Light Blue 9.14 m 25 mm
£67.06
1 box
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Add to basket
12850590
View Documents
Brady™
142305
Light Blue 15 m 50 mm
£84.74
1 box
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Add to basket
12800570
View Documents
Brady™
142284
Orange 15 m 13 mm
£47.32
1 box
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Add to basket
10762477
View Documents
Brady™
142285
Orange 15 m 25 mm
£67.06
1 box
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Add to basket
10785305
View Documents
Brady™
142286
Orange 15 m 50 mm
£84.74
1 box
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Add to basket
12810570
View Documents
Brady™
142299
Purple 15 m 13 mm
£47.32
1 box
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Add to basket
12820580
View Documents
Brady™
142300
Purple 15 m 25 mm
£67.06
1 box
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Add to basket
12830590
View Documents
Brady™
142301
Purple 15 m 50 mm
£84.74
1 box
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Add to basket
12820570
View Documents
Brady™
142278
Red 15 m 13 mm
£47.32
1 box
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Add to basket
12830580
View Documents
Brady™
142279
Red 15 m 25 mm
£67.06
1 box
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Add to basket
12830570
View Documents
Brady™
142269
White 15 m 13 mm £47.32
1 box
Qty Price Break Available View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Add to basket
12850580
View Documents
Brady™
142270
White 15 m 25 mm
£67.06
1 box
View alternative products
Save Up to 
»
Estimated Shipment: 19-08-2021
Add to basket
12860580
View Documents
Brady™
142273
Yellow 15 m 25 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Specifications
2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
Gloss
HandiMark Printer
HandiMark Indoor/Outdoor Tape
0.024 mm
Vinyl
Powder-coated
For facility and safety identification
60° 50 Gardner Units
-40°C to +82°C
Acrylic, Permanent, Cold Temperature